Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007413.1 | NP_001007414.1 | 2 - 3 | 437 | 74 | |
GenBank mRNA | BC085372.1 | - | 2 - 3 | 437 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |