Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017994.2 | NP_001017994.1 | 7 - 8 | 823 | 82 | |
XM_005156782.2 | XP_005156839.1 | 8 - 9 | 82 | |||
GenBank mRNA | AY394932.1 | - | 8 - 9 | 82 | ||
BC081633.1 | - | 7 - 8 | 823 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |