Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076632.1 | NP_001070100.1 | 7 - 8 | 506 | 97 | |
GenBank mRNA | BC124342.1 | - | 7 - 8 | 506 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |