Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtus1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077608.2 | NP_001071076.2 | 5 - 6 | 856 | 150 | |
XM_005157240.4 | XP_005157297.1 | 10 - 11 | 3144 | 150 | ||
XM_005157242.4 | XP_005157299.1 | 4 - 5 | 551 | 150 | ||
XM_017359019.2 | XP_017214508.1 | 10 - 11 | 3141 | 150 | ||
XM_021481039.1 | XP_021336714.1 | 7 - 8 | 1091 | 150 | ||
GenBank mRNA | BC125970.1 | - | 5 - 6 | 856 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |