Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 23 - 24 | 3402 | 91 | |
XM_005168796.4 | XP_005168853.1 | 24 - 25 | 6238 | 91 | ||
GenBank mRNA | BC066564.1 | - | 23 - 24 | 4284 | 91 | |
BC129198.1 | - | 23 - 24 | 3402 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |