Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083865.1 | NP_001077334.1 | 1 - 2 | 119 | 65 | |
NM_131049.1 | NP_571124.1 | 3 - 4 | 663 | 65 | ||
XM_005168429.4 | XP_005168486.1 | 3 - 4 | 652 | 65 | ||
XM_017359054.2 | XP_017214543.1 | 3 - 4 | 631 | 65 | ||
XM_021481386.1 | XP_021337061.1 | 2 - 3 | 593 | 65 | ||
GenBank mRNA | AF073289.1 | - | 3 - 4 | 663 | 65 | |
BC078280.1 | - | 2 - 3 | 268 | 65 | ||
BC155168.1 | - | 3 - 4 | 626 | 65 | ||
CU458994.1 | - | 1 - 2 | 119 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |