Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130960.1 | NP_571035.1 | 2 - 3 | 551 | 80 | |
GenBank mRNA | AF132444.1 | - | 2 - 3 | 534 | 80 | |
AF172089.1 | - | 2 - 3 | 551 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |