Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200193.1 | NP_956487.1 | 2 - 3 | 305 | 84 | |
XM_021468776.1 | XP_021324451.1 | 2 - 3 | 280 | 84 | ||
GenBank mRNA | BC045904.1 | - | 2 - 3 | 305 | 84 | |
BC164512.1 | - | 1 - 2 | 102 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |