Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200632.2 | NP_956926.1 | 5 - 6 | 621 | 73 | |
GenBank mRNA | BC057415.1 | - | 4 - 5 | 522 | 73 | |
EH509759.1 | - | 5 - 6 | 601 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |