Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc14aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201149.1 | NP_957443.1 | 10 - 11 | 1297 | 75 | |
XM_005163259.4 | XP_005163316.1 | 10 - 11 | 1359 | 75 | ||
GenBank mRNA | BC055188.1 | - | 10 - 11 | 1297 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |