Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201313.1 | NP_958470.1 | 1 - 2 | 247 | 77 | |
XM_005168471.4 | XP_005168528.1 | 1 - 2 | 255 | 77 | ||
GenBank mRNA | BC047809.1 | - | 1 - 2 | 247 | 77 | |
BC066630.1 | - | 1 - 2 | 216 | 77 | ||
BC165728.1 | - | 1 - 2 | 244 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |