Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207057.1 | NP_996940.1 | 11 - 12 | 1468 | 59 | |
XM_005161590.4 | XP_005161647.1 | 11 - 12 | 1426 | 59 | ||
GenBank mRNA | BC066393.1 | - | 11 - 12 | 1468 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |