Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213350.1 | NP_998515.1 | 2 - 3 | 194 | 83 | |
GenBank mRNA | BC054576.1 | - | 2 - 3 | 194 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |