Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nxph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002733.3 | NP_001002733.2 | 1 - 2 | 382 | 87 | |
GenBank mRNA | BC076424.1 | - | 1 - 2 | 386 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |