Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003878.2 | NP_001003878.2 | 2 - 3 | 249 | 98 | |
GenBank mRNA | AY648825.1 | - | 2 - 3 | 261 | 98 | |
BC095551.1 | - | 2 - 3 | 249 | 98 | ||
EH592323.1 | - | 2 - 3 | 525 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |