Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cratb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004594.2 | NP_001004594.2 | 11 - 12 | 1547 | 64 | |
GenBank mRNA | BC081604.1 | - | 11 - 12 | 1474 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |