Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012486.1 | NP_001012504.1 | 22 - 23 | 3121 | 137 | |
XM_009306437.3 | XP_009304712.1 | 22 - 23 | 3520 | 137 | ||
XM_009306438.3 | XP_009304713.1 | 21 - 22 | 3514 | 137 | ||
XM_009306439.3 | XP_009304714.1 | 22 - 23 | 3520 | 137 | ||
XM_009306440.3 | XP_009304715.1 | 22 - 23 | 3520 | 137 | ||
GenBank mRNA | AY772391.1 | - | 22 - 23 | 3121 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |