Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spopla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013447.1 | NP_001013465.1 | 4 - 5 | 468 | 136 | |
GenBank mRNA | BC090815.1 | - | 4 - 5 | 468 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |