Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013453.1 | NP_001013471.1 | 4 - 5 | 488 | 132 | |
XM_005165512.4 | XP_005165569.1 | 4 - 5 | 476 | 132 | ||
XM_021476140.1 | XP_021331815.1 | 4 - 5 | 557 | 132 | ||
GenBank mRNA | BC091559.1 | - | 4 - 5 | 488 | 132 | |
DQ294346.1 | - | 4 - 5 | 468 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |