Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017728.2 | NP_001017728.2 | 3 - 4 | 490 | 90 | |
GenBank mRNA | BC093227.1 | - | 3 - 4 | 483 | 90 | |
EH551524.1 | - | 3 - 4 | 529 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |