Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017893.1 | NP_001017893.1 | 2 - 3 | 130 | 132 | |
GenBank mRNA | BC092953.1 | - | 2 - 3 | 130 | 132 | |
BC164438.1 | - | 2 - 3 | 113 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |