Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018140.2 | NP_001018150.1 | 11 - 12 | 1710 | 67 | |
NM_001328167.1 | NP_001315096.1 | 16 - 17 | 2259 | 67 | ||
XM_005158845.4 | XP_005158902.1 | 11 - 12 | 1624 | 67 | ||
XM_009293199.3 | XP_009291474.1 | 15 - 16 | 2146 | 67 | ||
XM_009293200.3 | XP_009291475.1 | 11 - 12 | 1717 | 67 | ||
XM_021467161.1 | XP_021322836.1 | 16 - 17 | 2399 | 67 | ||
XM_021467162.1 | XP_021322837.1 | 16 - 17 | 2309 | 67 | ||
XM_021467163.1 | XP_021322838.1 | 16 - 17 | 2310 | 67 | ||
GenBank mRNA | AB196917.1 | - | 11 - 12 | 1712 | 67 | |
BC162582.1 | - | 11 - 12 | 1622 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |