Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020612.1 | NP_001018448.1 | 3 - 4 | 358 | 96 | |
GenBank mRNA | BC095337.1 | - | 3 - 4 | 358 | 96 | |
BC164152.1 | - | 3 - 4 | 323 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |