Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025551.1 | NP_001020722.1 | 7 - 8 | 1989 | 67 | |
XM_005160586.3 | XP_005160643.1 | 8 - 9 | 2558 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |