Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vgll2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033095.1 | NP_001028267.1 | 3 - 4 | 1012 | 73 | |
GenBank mRNA | BC100003.1 | - | 3 - 4 | 1012 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |