Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnikb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037676.2 | NP_001032765.2 | 32 - 33 | 4184 | 132 | |
XM_005162778.4 | XP_005162835.1 | 32 - 33 | 4544 | 132 | ||
XM_005162779.4 | XP_005162836.1 | 31 - 32 | 4189 | 132 | ||
XM_005162780.4 | XP_005162837.1 | 30 - 31 | 4102 | 132 | ||
XM_021470183.1 | XP_021325858.1 | 34 - 35 | 4577 | 132 | ||
XM_021470184.1 | XP_021325859.1 | 33 - 34 | 4490 | 132 | ||
XM_021470185.1 | XP_021325860.1 | 33 - 34 | 4430 | 132 | ||
XM_021470186.1 | XP_021325861.1 | 33 - 34 | 4418 | 132 | ||
XM_021470187.1 | XP_021325862.1 | 33 - 34 | 4222 | 132 | ||
XM_021470188.1 | XP_021325863.1 | 32 - 33 | 4135 | 132 | ||
GenBank mRNA | BC108020.1 | - | 13 - 14 | 1793 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |