Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: illr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040039.1 | NP_001035128.1 | 2 - 3 | 271 | 81 | |
NM_001044742.1 | NP_001038207.1 | 2 - 3 | 244 | 81 | ||
XM_021468134.1 | XP_021323809.1 | 2 - 3 | 298 | 81 | ||
GenBank mRNA | AY986756.1 | - | 2 - 3 | 271 | 81 | |
AY986757.1 | - | 2 - 3 | 244 | 81 | ||
BC163143.1 | - | 2 - 3 | 244 | 81 | ||
BC163144.1 | - | 2 - 3 | 244 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |