Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110450.1 | NP_001103920.1 | 3 - 4 | 349 | 100 | |
GenBank mRNA | BC134941.1 | - | 3 - 4 | 349 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |