Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131523.1 | NP_571598.1 | 8 - 9 | 674 | 96 | |
GenBank mRNA | AF229154.1 | - | 8 - 9 | 674 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |