Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173263.1 | NP_775370.1 | 6 - 7 | 662 | 74 | |
GenBank mRNA | AY099512.1 | - | 6 - 7 | 662 | 74 | |
AY394968.1 | - | 6 - 7 | 675 | 74 | ||
BC058315.1 | - | 6 - 7 | 658 | 74 | ||
BC066518.1 | - | 5 - 6 | 648 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |