Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200404.1 | NP_956698.1 | 1 - 2 | 379 | 74 | |
GenBank mRNA | BC054577.1 | - | 1 - 2 | 379 | 74 | |
BC153629.1 | - | 1 - 2 | 385 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |