Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200718.1 | NP_957012.1 | 10 - 11 | 759 | 69 | |
GenBank mRNA | BC059479.1 | - | 10 - 11 | 759 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |