Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pafah1b1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201346.1 | NP_958503.1 | 8 - 9 | 1200 | 67 | |
XM_021468786.1 | XP_021324461.1 | 8 - 9 | 1124 | 67 | ||
GenBank mRNA | BC044530.1 | - | 8 - 9 | 1228 | 67 | |
DQ141216.1 | - | 7 - 8 | 970 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |