Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ywhaqa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201513.1 | NP_958921.1 | 5 - 6 | 859 | 104 | |
GenBank mRNA | BC048068.1 | - | 5 - 6 | 859 | 104 | |
BC066409.1 | - | 4 - 5 | 834 | 104 | ||
BC154307.1 | - | 5 - 6 | 855 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |