Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001403.2 | NP_001001403.2 | 18 - 19 | 2664 | 55 | |
GenBank mRNA | BC055391.1 | - | 18 - 19 | 2664 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |