Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002351.1 | NP_001002351.1 | 7 - 8 | 1267 | 80 | |
GenBank mRNA | BC075908.1 | - | 7 - 8 | 1267 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |