Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007415.1 | NP_001007416.1 | 3 - 4 | 413 | 75 | |
XM_005171163.4 | XP_005171220.1 | 3 - 4 | 274 | 75 | ||
XM_021480037.1 | XP_021335712.1 | 3 - 4 | 392 | 75 | ||
GenBank mRNA | BC085370.1 | - | 3 - 4 | 413 | 75 | |
BC165850.1 | - | 2 - 3 | 192 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |