Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtac1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080178.1 | NP_001073647.1 | 10 - 11 | 1329 | 67 | |
XM_005160175.4 | XP_005160232.1 | 10 - 11 | 1411 | 67 | ||
XM_021475056.1 | XP_021330731.1 | 10 - 11 | 1412 | 67 | ||
XM_021475062.1 | XP_021330737.1 | 10 - 11 | 1380 | 67 | ||
XM_021475068.1 | XP_021330743.1 | 10 - 11 | 1410 | 67 | ||
GenBank mRNA | DQ351708.1 | - | 10 - 11 | 1329 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |