Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtac1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080178.1 | NP_001073647.1 | 7 - 8 | 1008 | 68 | |
XM_005160175.4 | XP_005160232.1 | 7 - 8 | 1090 | 68 | ||
XM_021475056.1 | XP_021330731.1 | 7 - 8 | 1091 | 68 | ||
XM_021475062.1 | XP_021330737.1 | 7 - 8 | 1059 | 68 | ||
XM_021475068.1 | XP_021330743.1 | 7 - 8 | 1089 | 68 | ||
GenBank mRNA | BC146702.1 | - | 7 - 8 | 1029 | 68 | |
DQ351708.1 | - | 7 - 8 | 1008 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |