Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099983.1 | NP_001093453.1 | 1 | 449 | 66 | |
GenBank mRNA | CR930857.1 | - | 1 | 449 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |