Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131565.1 | NP_571640.1 | 3 - 4 | 90 | 72 | |
XM_005163151.3 | XP_005163208.3 | 4 - 5 | 230 | 72 | ||
GenBank mRNA | AF180889.1 | - | 3 - 4 | 90 | 72 | |
BC053304.1 | - | 4 - 5 | 113 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |