Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173225.1 | NP_775332.1 | 14 - 15 | 1798 | 74 | |
GenBank mRNA | AF462310.1 | - | 14 - 15 | 1798 | 74 | |
BC085524.1 | - | 14 - 15 | 1796 | 74 | ||
BC165677.1 | - | 14 - 15 | 1748 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |