Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199428.2 | NP_955460.2 | 7 - 8 | 679 | 182 | |
GenBank mRNA | BC053240.1 | - | 7 - 8 | 688 | 194 | |
BC153486.1 | - | 7 - 8 | 588 | 188 | ||
BC164639.1 | - | 7 - 8 | 589 | 194 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |