Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200067.1 | NP_956361.1 | 3 - 4 | 732 | 81 | |
GenBank mRNA | BC049516.1 | - | 3 - 4 | 732 | 81 | |
BC075766.1 | - | 3 - 4 | 732 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |