Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200256.1 | NP_956550.1 | 4 - 5 | 795 | 94 | |
GenBank mRNA | BC048057.1 | - | 4 - 5 | 795 | 94 | |
BC164127.1 | - | 4 - 5 | 623 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |