Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200727.1 | NP_957021.1 | 3 - 4 | 484 | 91 | |
GenBank mRNA | BC059501.1 | - | 3 - 4 | 484 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |