Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214739.1 | NP_999904.1 | 2 - 3 | 177 | 109 | |
GenBank mRNA | AB264293.1 | - | 2 - 3 | 96 | 109 | |
BC067189.1 | - | 2 - 3 | 177 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |