Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214739.1 | NP_999904.1 | 3 - 4 | 281 | 112 | |
GenBank mRNA | BC067189.1 | - | 3 - 4 | 281 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |