Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asic1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214791.1 | NP_999956.1 | 8 - 9 | 1572 | 63 | |
XM_005161666.4 | XP_005161723.1 | 8 - 9 | 1856 | 63 | ||
XM_005161667.4 | XP_005161724.1 | 9 - 10 | 2308 | 63 | ||
GenBank mRNA | AJ609615.1 | - | 8 - 9 | 1572 | 63 | |
BC162362.1 | - | 8 - 9 | 1541 | 63 | ||
BC162368.1 | - | 8 - 9 | 1541 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |