Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr0b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082947.1 | NP_001076416.1 | 2 | 753 | 138 | |
GenBank mRNA | BC133942.1 | - | 2 | 753 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |